Learn More
Invitrogen™ MBTPS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109948
Description
Immunogen sequence: ITQTFKDQGL EVLKQETAVV ENVPILGLYQ IPAEGGGRIV LYGDSNCLDD SHRQKDCFWL LDALLQYTSY GVTPPSLSHS GNRQRPPSGA GSVTPERMEG NHLHRYSKVL EAHLGDPKPR PLPACPRLSW AKPQPLNETA PSNLWKHQKL LSIDLDKVVL PNFRSNRPQV RPLSPGESGA WDIPGGIMPG RYNQEVGQTI PVFAFLGAMV VLAFFVVQIN KAKSRPKRRK PRVKRPQLMQ QVHPPKTPSV.
This gene encodes a member of the subtilisin-like proprotein convertase family, which includes proteases that process protein and peptide precursors trafficking through regulated or constitutive branches of the secretory pathway. The encoded protein undergoes an initial autocatalytic processing event in the ER to generate a heterodimer which exits the ER and sorts to the cis/medial-Golgi where a second autocatalytic event takes place and the catalytic activity is acquired. It encodes a type 1 membrane bound protease which is ubiquitously expressed and regulates cholesterol or lipid homeostasis via cleavage of substrates at non-basic residues. Mutations in this gene may be associated with lysosomal dysfunction.
Specifications
MBTPS1 | |
Polyclonal | |
Unconjugated | |
MBTPS1 | |
0610038M03Rik; AV003995; endopeptidase S1P; KIAA0091; MBTPS1; membrane bound transcription factor peptidase, site 1; membrane-bound transcription factor peptidase, site 1; membrane-bound transcription factor protease, site 1; membrane-bound transcription factor site-1 protease; mKIAA0091; PCSK8; proprotein convertase subtilisin/kexin type 8; S1p; S1P endopeptidase; site-1 protease; Ski1; SKI-1; Sterol-regulated luminal protease; subtilisin kexin isozyme 1; subtilisin/kexin isozyme 1; subtilisin/kexin isozyme SKI-1; subtilisin/kexin isozyme-1; Subtilisin/kexin-isozyme 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56453, 8720 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.21 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q14703, Q9WTZ2 | |
MBTPS1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 803-1052 of human MBTPS1 (NP_003782.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.