Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBTPS2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | MBTPS2 |
---|---|
Concentration | 0.05mg/mL |
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Description
MBTPS2 Polyclonal specifically detects MBTPS2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MBTPS2 | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Human | |
O43462 | |
51360 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:EVAEDSPAIGPRGLFVGDLVTHLQDCPVTNVQDWNECLDTIAYEPQIGYCISASTLQQLSFPVRAYKRLDGSTECCNNHSLTDVCFSYRNNFNKRLHTCLPARKAVEATQVCRTNKDCKKSSSSSFCIIPSLETHTR | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.05mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
EC 3.4.24.85, Endopeptidase S2P, FLJ32174, membrane-bound transcription factor peptidase, site 2, membrane-bound transcription factor protease, site 2, membrane-bound transcription factor site-2 protease, S2PIFAP, SREBPs intramembrane protease, Sterol regulatory element-binding proteins intramembrane protease | |
MBTPS2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title