Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBTPS2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548486
Description
Predicted to react with bovine and mouse based on sequence homology.
This gene encodes a intramembrane zinc metalloprotease, which is essential in development. This protease functions in the signal protein activation involved in sterol control of transcription and the ER stress response. Mutations in this gene have been associated with ichthyosis follicularis with atrichia and photophobia (IFAP syndrome); IFAP syndrome has been quantitatively linked to a reduction in cholesterol homeostasis and ER stress response.[provided by RefSeq, Aug 2009].
Specifications
MBTPS2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
O43462 | |
MBTPS2 | |
KLH conjugated synthetic peptide between 15-44 amino acids from the N-terminal region of human MBTPS2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
MBTPS2 | |
Membrane-bound transcription factor site-2 protease, Endopeptidase S2P, Sterol regulatory element-binding proteins intramembrane protease, SREBPs intramembrane protease, MBTPS2, S2P | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51360 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction