Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBTPS2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2852610.1ML
Description
MBTPS2 Polyclonal specifically detects MBTPS2 in Human samples. It is validated for Western Blot.Specifications
MBTPS2 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
51360 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
EC 3.4.24.85, Endopeptidase S2P, FLJ32174, membrane-bound transcription factor peptidase, site 2, membrane-bound transcription factor protease, site 2, membrane-bound transcription factor site-2 protease, S2PIFAP, SREBPs intramembrane protease, Sterol regulatory element-binding proteins intramembrane protease | |
The immunogen is a synthetic peptide directed towards the N-terminal region of human MBTPS2 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction