Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MC-1R/MC1 Receptor Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983219
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3219 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3219 Supplier Bioss Supplier No. BS1419R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MC1R, a Melanocortin Receptor, mediates the effects of melanocyte-stimulating hormone (MSH) and adrenocorticotropic hormone (ACTH). MSH and its receptor, MC1R, are key regulators of melanogenesis. They control the relative proportions of red pheomelanin and black photoprotective eumelanin in the skin by increasing the production of eumelanin. Non-functional, truncated forms of the receptor lead to lighter coat color in animals. In contrast, constitutively active receptors lead to darker color.

Specifications

Antigen MC-1R/MC1 Receptor
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Mc1r
Gene Accession No. Q01726
Gene Alias alpha melanocyte stimulating hormone receptor; CMM5; e; extension recessive yellow; MC1 receptor; MC1R; MC1-R; melanocortin 1 receptor; melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor); Melanocortin receptor 1; melanocortin-1 receptor; Melanocyte-stimulating hormone receptor; melanotropin receptor; MSHR; Msh-r; Mshra; SHEP2; Tob; tobacco darkening
Gene Symbols Mc1r
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse MC-1R .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4157
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.