Learn More
Invitrogen™ MC1R Polyclonal Antibody

Description
MC1R Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA
Specifications
Specifications
| Antigen | MC1R |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | Mc1r |
| Gene Accession No. | Q01726 |
| Gene Alias | alpha melanocyte stimulating hormone receptor; CMM5; e; extension recessive yellow; MC1 receptor; MC1R; MC1-R; melanocortin 1 receptor; melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor); melanocortin receptor 1; melanocortin-1 receptor; melanocyte-stimulating hormone receptor; melanotropin receptor; MSHR; MSH-R; Mshra; SHEP2; Tob; tobacco darkening |
| Gene Symbols | Mc1r |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.