Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MC1R Recombinant Rabbit Monoclonal Antibody (HL1471)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550315
Description
MC1R Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
MC1R, a Melanocortin Receptor, mediates the effects of melanocyte-stimulating hormone (MSH) and adrenocorticotropic hormone (ACTH). MSH and its receptor, MC1R, are key regulators of melanogenesis. They control the relative proportions of red pheomelanin and black photoprotective eumelanin in the skin by increasing the production of eumelanin. Non-functional, truncated forms of the receptor lead to lighter coat color in animals. In contrast, constitutively active receptors lead to darker color.
Specifications
MC1R | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q01726, Q01727 | |
Mc1r | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human MC1 Receptor. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
HL1471 | |
Unconjugated | |
Mc1r | |
alpha melanocyte stimulating hormone receptor; CMM5; e; extension recessive yellow; MC1 receptor; MC1R; MC1-R; melanocortin 1 receptor; melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor); melanocortin receptor 1; melanocortin-1 receptor; melanocyte-stimulating hormone receptor; melanotropin receptor; MSHR; MSH-R; Mshra; SHEP2; Tob; tobacco darkening | |
Rabbit | |
Protein A | |
RUO | |
102552838, 17199, 4157 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction