Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MC2R Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550517
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550517 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550517 Supplier Invitrogen™ Supplier No. PA550517
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total MC2R protein.

MC2R encodes one member of the five-member G-protein associated melanocortin receptor family. Melanocortins are peptides derived from pro-opiomelanocortin. MC2R is selectively activated by adrenocorticotropic hormone, whereas the other four melanocortin receptors recognize a variety of melanocortin ligands. Mutations in MC2R can result in familial glucocorticoid deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MC2R
Applications Western Blot
Classification Polyclonal
Concentration 2.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MC2R
Gene Accession No. Q01718
Gene Alias ACTH receptor; ACTHR; ACTH-R; Adrenocorticotropic hormone receptor; adrenocorticotropin receptor; corticotropin receptor; MC2 receptor; MC2R; MC2-R; melanocortin 2 receptor; melanocortin 2 receptor (adrenocorticotropic hormone); melanocortin receptor 2; MGC125798
Gene Symbols MC2R
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the N terminal of human Melanocortin 2 receptor (adrenocorticotropic hormone).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4158
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.