Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MC3R Monoclonal Antibody (345404)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA524082
Description
This antibody stains mouse Melanocortin 3R/MC3R-transfected cells but not irrelevant transfectants. Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a G-protein-coupled receptor for melanocyte-stimulating hormone and adrenocorticotropic hormone that is expressed in tissues other than the adrenal cortex and melanocytes. This gene maps to the same region as the locus for benign neonatal epilepsy. Mice deficient for this gene have increased fat mass despite decreased food intake, suggesting a role for this gene product in the regulation of energy homeostasis. Mutations in this gene are associated with a susceptibility to obesity in humans.
Specifications
MC3R | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P33033 | |
MC3R | |
Y3 rat myeloid cell line transfeced with mouse Melanocortin 3R/MC3R. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
345404 | |
Unconjugated | |
MC3R | |
BMIQ9; MC3; MC3 receptor; MC3R; MC3-R; melanocortin 3 receptor; melanocortin receptor 3; OB20; obesity quantitative trait locus; OQTL | |
Rat | |
Protein A/G | |
RUO | |
17201 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction