Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MC3R Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588196
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588196 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588196 Supplier Invitrogen™ Supplier No. PA588196
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNASCCLPSV QPTLPNGSEH LQAPFFSNQS SSAFCEQVFI KPEVFLSLGI VSLLENILVI LAVVRNGNLH SPMYFFLCSL AVADMLVSVS NALETIMIAI VHSDYLTFED QFIQHMDNIF; Positive Samples: K-562; Cellular Location: Cell membrane, Multi-pass membrane protein.

This gene encodes a G-protein-coupled receptor for melanocyte-stimulating hormone and adrenocorticotropic hormone that is expressed in tissues other than the adrenal cortex and melanocytes. This gene maps to the same region as the locus for benign neonatal epilepsy. Mice deficient for this gene have increased fat mass despite decreased food intake, suggesting a role for this gene product in the regulation of energy homeostasis. Mutations in this gene are associated with a susceptibility to obesity in humans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MC3R
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MC3R
Gene Accession No. P32244, P33033, P41968
Gene Alias BMIQ9; MC3; MC3 receptor; MC3R; MC3-R; melanocortin 3 receptor; melanocortin receptor 3; OB20; obesity quantitative trait locus; OQTL
Gene Symbols MC3R
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human MC3R (NP_0639413).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17201, 29310, 4159
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.