Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MC5R Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533928
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533928 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533928 Supplier Thermo Scientific Supplier No. PA533928
Add to Cart
Add to Cart

MC5R Polyclonal Antibody

MC5R Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen MC5R
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene MC5R
Gene Accession No. P33032
Gene Alias H MC2; MC2; MC-2; MC5; MC5 receptor; MC5R; MC5-R; melanocortin 5 receptor; melanocortin receptor 5; melanocortin-5 receptor
Gene Symbols MC5R
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from N-terminal extracellular domain of human MC5 Receptor.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4161
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.