Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MC5R Recombinant Rabbit Monoclonal Antibody (JE61-09)
GREENER_CHOICE

Catalog No. PIMA544865
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544865 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544865 Supplier Invitrogen™ Supplier No. MA544865
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

MC5R Recombinant Monoclonal Antibody for Western Blot, IHC (P)

MC5R, a Melanocortin Receptor, mediates the effects of melanocyte-stimulating hormone (MSH) and adrenocorticotropic hormone (ACTH), and activates adenylate cyclase. This receptor controls multiple functions in the brain and in peripheral tissues, such as thermoregulation, immunomodulation, appetite control, and sexual behavior. Melanocortin 5 receptor has been reported to be expressed in a variety of tissues, especially in pituitary, exocrine glands, reproductive organs, lung, kidney, and esophagus. No human ESTs have been isolated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MC5R
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE61-09
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MC5R
Gene Accession No. P33032, P41149
Gene Alias H MC2; MC2; MC-2; MC5; MC5 receptor; MC5R; MC5-R; melanocortin 5 receptor; melanocortin receptor 5; melanocortin-5 receptor
Gene Symbols MC5R
Host Species Rabbit
Immunogen Synthetic peptide within human MC-2 aa 1-50/325.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17203, 4161
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.