Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCAF1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100438JF549
Description
MCAF1 Polyclonal antibody specifically detects MCAF1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ImmunoblotSpecifications
MCAF1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting | |
activating transcription factor 7 interacting protein, activating transcription factor 7-interacting protein 1, ATF7-interacting protein, ATFa-associated modulator, ATF-interacting protein, ATF-IP, FLJ10139, FLJ10688, hAM, MBD1-containing chromatin associated factor, MCAF1AM, MCAFMBD1-containing chromatin-associated factor 1, MMCP 6, MMCP6, MMCP-6, P621 | |
A synthetic peptide that maps to a region between residues 25 and 100 of human MBD1-containing Chromatin Associated Factor using the numbering given in entry AAQ92978.1 (GeneID 55729) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoblot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55729 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction