Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCAF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559456
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559456 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559456 Supplier Invitrogen™ Supplier No. PA559456
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVESPNLTTP ITSNPTDTRK ITSGNSSNSP NAEVMAVQKK LDSIIDLTKE GLSNCNTESP VSPLESHSKA ASNSKETTPL AQNAVQVPES FEHLPPLPEP PAPL Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 40%.

Recruiter that couples transcriptional factors to general transcription apparatus and thereby modulates transcription regulation and chromatin formation. Can both act as an activator or a repressor depending on the context. Mediates MBD1-dependent transcriptional repression, probably by recruiting complexes containing SETDB1. The complex formed with MBD1 and SETDB1 represses transcription and probably couples DNA methylation and histone H3 'Lys-9' trimethylation (H3K9me3) activity (Probable). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCAF2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ATF7IP2
Gene Accession No. Q5U623
Gene Alias 4930558K11Rik; activating transcription factor 7 interacting protein 2; activating transcription factor 7-interacting protein 2; ATF7-interacting protein 2; ATF7IP2; BC018510; Get-1; MBD1-containing chromatin associated factor 2; MBD1-containing chromatin-associated factor 2; Mcaf2; Protein similar to MCAF2; PSM2
Gene Symbols ATF7IP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MCAF2. Recombinant protein control fragment (Product #RP-98717).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80063
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.