Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCAK Monoclonal Antibody (OTI1C5)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525647

Catalog No. PIMA525647


Only null left
Add to Cart

Description

Description

MCAK Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC), which consists of 60-100 proteins. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion while larger molecules are transported by an active process. The protein encoded by this gene belongs to the importin alpha family, and is involved in nuclear protein import.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MCAK
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q62909, Q922S8, Q99661
KIF2C
Full length human recombinant protein of KIF2C produced in HEK293T cell.
100 μL
Primary
Human, Mouse, Rat, Monkey
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI1C5
Unconjugated
KIF2C
4930402F02Rik; CT139; ESTM5; hypothetical protein; KIF2C; kinesin family member 2C; kinesin-like 6; kinesin-like protein 6; Kinesin-like protein KIF2C; kinesin-related protein 2; KNSL6; KRP2; MCAK; Mitotic centromere-associated kinesin; QtsA-16015; RP11-269F19.1; testis tissue sperm-binding protein Li 68n; X83316
Mouse
Affinity Chromatography
RUO
11004, 171529, 73804
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.