Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCAK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140304
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140304 Supplier Invitrogen™ Supplier No. PA5140304
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: MDSSLQARLF PGLAIKIQRS NGLIHSANVR TVNLEKSCVS VEWAEGGATK GKEIDFDDVA AINPELLQLL PLHPKDNLPL QENVTIQK Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 81%.

The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC), which consists of 60-100 proteins. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion while larger molecules are transported by an active process. The protein encoded by this gene belongs to the importin alpha family, and is involved in nuclear protein import.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCAK
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIF2C
Gene Accession No. Q99661
Gene Alias 4930402F02Rik; CT139; ESTM5; hypothetical protein; KIF2C; kinesin family member 2C; kinesin-like 6; kinesin-like protein 6; Kinesin-like protein KIF2C; kinesin-related protein 2; KNSL6; KRP2; MCAK; Mitotic centromere-associated kinesin; QtsA-16015; RP11-269F19.1; testis tissue sperm-binding protein Li 68n; X83316
Gene Symbols KIF2C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIF2C. Recombinant protein control fragment (Product #RP-109690).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11004
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.