Learn More
MCAK Rabbit anti-Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300921A
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Rat
The protein encoded by this gene is a member of kinesin-like protein family. Proteins of this family are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. This protein is important for anaphase chromosome segregation and may be required to coordinate the onset of sister centromere separation.Specifications
| MCAK | |
| Polyclonal | |
| Unconjugated | |
| Kif2c | |
| RP11-269F19.1, KNSL6, MCAK, kinesin-like 6; kinesin-like protein 6; kinesin-like protein KIF2C; mitotic centromere-associated kinesin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 73804 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q922S8 | |
| Kif2c | |
| Between 50 and 100 | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.