Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCAM/CD146 Antibody (733216) [DyLight 350], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB7718D
Description
MCAM/CD146 Monoclonal antibody specifically detects MCAM/CD146 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
MCAM/CD146 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
733216 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18Gicerin | |
Mouse myeloma cell line NS0-derived recombinant mouse MCAM/CD146, Met1-Val563, Accession # Q8R2Y2 | |
0.1 mL | |
Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
4162 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction