Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MCAM/CD146 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB932R100UG
This item is not returnable.
View return policy
Description
MCAM/CD146 Monoclonal specifically detects MCAM/CD146 in Human samples. It is validated for Flow Cytometry.Specifications
MCAM/CD146 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAA20824 | |
MCAM | |
Mouse myeloma cell line NS0-derived recombinant human MCAM Val24-Gly559 Accession # AAA20824 | |
100 μg | |
Primary | |
Detects human MCAM in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ALCAM, rhBCAM, rhEpCAM, rhNCAM-L1, recombinant mouse (rm) OCAM, or rmMAdCAM-1 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Flow Cytometry | |
128018 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, L-Gicerin, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18, MUC18Gicerin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4162 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction