Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCAM/CD146 Rabbit anti-Human, Clone: 5, Novus Biologicals™

Rabbit Monoclonal Antibody
$524.00
Specifications
Antigen | MCAM/CD146 |
---|---|
Clone | 5 |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:2000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
MCAM/CD146 Monoclonal specifically detects MCAM/CD146 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MCAM/CD146 | |
Western Blot 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18Gicerin | |
Recombinant Human MCAM/CD146 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
5 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
0.2 um filtered solution in PBS | |
4162 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title