Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCAR1/MCAR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144057
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144057 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144057 Supplier Invitrogen™ Supplier No. PA5144057
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, monkey COS-7 whole cell, mouse NIH/3T3 whole cell. IHC: human laryngeal squamous cell carcinomas tissue, human serous adenocarcinoma of ovary tissue. ICC/IF: Hela cell. Flow: U251 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

MCAR1 enables NAD transmembrane transporter activity. Involved in mitochondrial NAD transmembrane transport. Located in mitochondrion. Is active in mitochondrial inner membrane. MCAR2 is a mitochondrial membrane carrier protein that mediates the import of NAD(+) into mitochondria. Compared to MCAR1, MCAR2-mediated transport is not essential for the import of NAD(+) in mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCAR1/MCAR2
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene SLC25A52
Gene Accession No. A0A2J8MLX0, A0A7N9CMS4, Q3SY17, Q5HZI9, Q9H1U9
Gene Alias MCART1; MCART2; SLC25A51; SLC25A52
Gene Symbols SLC25A51, SLC25A52
Host Species Rabbit
Immunogen E.coli-derived human SLC25A51/52 recombinant protein (Position: M1-E291).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102135726, 147407, 230125, 468613, 92014
Target Species Human, Mouse, Monkey
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.