Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCAR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563542 Supplier Invitrogen™ Supplier No. PA563542
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MMDSEAHEKR PPILTSSKQD ISPHITNVGE MKHYLC Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 69%.

SLC25A51 gene ontology annotations related to this gene include transmembrane transporter activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCAR1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC25A51
Gene Accession No. Q9H1U9
Gene Alias 9130208E07Rik; CG7943; D130005A03Rik; Gm138; Mcart1; mitochondrial carrier triple repeat 1; mitochondrial carrier triple repeat protein 1; mitochondrial carrier triple repeat protein 1-like; SLC25A51; Solute carrier family 25 member 51; solute carrier family 25, member 51
Gene Symbols SLC25A51
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MCAR1. Recombinant protein control fragment (Product #RP-104517).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92014
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.