Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCART1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS24079R

Description
SLC25A51 gene ontology annotations related to this gene include transmembrane transporter activity.Specifications
MCART1 | |
Polyclonal | |
Unconjugated | |
SLC25A51 | |
9130208E07Rik; CG7943; D130005A03Rik; Gm138; Mcart1; mitochondrial carrier triple repeat 1; Mitochondrial carrier triple repeat protein 1; SLC25A51; Solute carrier family 25 member 51; solute carrier family 25, member 51 | |
Rabbit | |
Protein A | |
RUO | |
92014 | |
-20°C | |
Liquid |
Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9H1U9 | |
SLC25A51 | |
KLH conjugated synthetic peptide derived from mouse MCART1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction