Learn More
Invitrogen™ MCAT Monoclonal Antibody (OTI2B7)

Description
MCAT Monoclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | MCAT |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI2B7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MCAT |
| Gene Accession No. | Q8IVS2 |
| Gene Alias | [Acyl-carrier-protein] malonyltransferase; [acyl-carrier-protein] S-malonyltransferase; AI225907; BC025519; fabD; FASN2C; malonyl CoA:ACP acyltransferase (mitochondrial); malonyl CoA-acyl carrier protein transacylase, mitochondrial; malonyl-CoA:acyl carrier protein transacylase, mitochondrial; malonyl-CoA-acyl carrier protein transacylase; malonyl-CoA-acyl carrier protein transacylase, mitochondrial; Mcat; MCT; mitochondrial malonyl CoA:ACP acyltransferase; Mitochondrial malonyltransferase; Mt; NET62; RGD1306698 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.