Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590735
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590735 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590735 Supplier Invitrogen™ Supplier No. PA590735
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGMLSVLGQP QSKFNFACLE AREHCKSLGI ENPVCEVSNY LFPDCRVISG HQEALRFLQK NSSKFHFRRT RMLPVSGAFH TRLMEPAVEP LTQALKAVDI KKPLVSVYSN VHAHRYRHPG HIHKLLAQQL VSPVKWEQTM HAIYERKKGR GFPQTFEVGP GRQLGAILKS CNMQAWKSYS AVDVLQTLEH VDLDPQEPPR; Positive Samples: LO2; Cellular Location: Mitochondrion.

Catalyzes the transfer of a malonyl moiety from malonyl-CoA to the free thiol group of the phosphopantetheine arm of the mitochondrial ACP protein (NDUFAB1). This suggests the existence of the biosynthesis of fatty acids in mitochondrias.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCAT
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MCAT
Gene Accession No. Q8IVS2
Gene Alias [Acyl-carrier-protein] malonyltransferase; [acyl-carrier-protein] S-malonyltransferase; AI225907; BC025519; fabD; FASN2C; malonyl CoA:ACP acyltransferase (mitochondrial); malonyl CoA-acyl carrier protein transacylase, mitochondrial; malonyl-CoA:acyl carrier protein transacylase, mitochondrial; malonyl-CoA-acyl carrier protein transacylase; malonyl-CoA-acyl carrier protein transacylase, mitochondrial; Mcat; MCT; mitochondrial malonyl CoA:ACP acyltransferase; Mitochondrial malonyltransferase; Mt; NET62; RGD1306698
Gene Symbols MCAT
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 191-390 of human MCAT (NP_7757383).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27349
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.