Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557283
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557283 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557283 Supplier Invitrogen™ Supplier No. PA557283
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HKLLAQQLVS PVKWEQTMHA IYERKKGRGF PQTFEVGPGR QLGAILKSCN MQAWKSYSAV DVLQTLEHVD LDPQEP Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 74%.

Catalyzes the transfer of a malonyl moiety from malonyl-CoA to the free thiol group of the phosphopantetheine arm of the mitochondrial ACP protein (NDUFAB1). This suggests the existence of the biosynthesis of fatty acids in mitochondrias.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCAT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MCAT
Gene Accession No. Q8IVS2
Gene Alias [Acyl-carrier-protein] malonyltransferase; [acyl-carrier-protein] S-malonyltransferase; AI225907; BC025519; fabD; FASN2C; malonyl CoA:ACP acyltransferase (mitochondrial); malonyl CoA-acyl carrier protein transacylase, mitochondrial; malonyl-CoA:acyl carrier protein transacylase, mitochondrial; malonyl-CoA-acyl carrier protein transacylase; malonyl-CoA-acyl carrier protein transacylase, mitochondrial; Mcat; MCT; mitochondrial malonyl CoA:ACP acyltransferase; Mitochondrial malonyltransferase; Mt; NET62; RGD1306698
Gene Symbols MCAT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MCAT. Recombinant protein control fragment (Product #RP-95356).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27349
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.