Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557933
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557933 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557933 Supplier Invitrogen™ Supplier No. PA557933
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VQELVSALER LTKSSEIRHQ QSAEFVNDLK RANSNLVAAY EKAKKKHQNK LKKLESQMMA MVERHETQVR MLKQRIALLE EENSRPHTNE TS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

This gene is a candidate colorectal tumor suppressor gene that is thought to negatively regulate cell cycle progression. The orthologous gene in the mouse expresses a phosphoprotein associated with the plasma membrane and membrane organelles, and overexpression of the mouse protein inhibits entry into S phase. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCC
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mcc
Gene Accession No. P23508
Gene Alias colorectal mutant cancer protein; D18Ertd451e; E330037C19; MCC; MCC1; mutated in colorectal cancers; Protein MCC
Gene Symbols Mcc
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MCC. Recombinant protein control fragment (Product #RP-104125).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4163
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.