Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MCC Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557934
Description
Immunogen sequence: ALASLKGDIV ELNKRLQQTE RERDLLEKKL AKAQCEQSHL MREHEDVQER TTLRYEERIT ELHSVIAELN KKIDRLQGTT IREEDEYSEL RSEL Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.
This gene is a candidate colorectal tumor suppressor gene that is thought to negatively regulate cell cycle progression. The orthologous gene in the mouse expresses a phosphoprotein associated with the plasma membrane and membrane organelles, and overexpression of the mouse protein inhibits entry into S phase. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| MCC | |
| Polyclonal | |
| Unconjugated | |
| Mcc | |
| colorectal mutant cancer protein; D18Ertd451e; E330037C19; MCC; MCC1; mutated in colorectal cancers; Protein MCC | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 307449, 328949, 4163 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P23508 | |
| Mcc | |
| Recombinant protein corresponding to Human MCC. Recombinant protein control fragment (Product #RP-96936). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction