Learn More
Invitrogen™ MCCC2 Monoclonal Antibody (2B3)

Description
Peptide Sequence: AYSPRFLYIW PNARISVMGG EQAANVLATI TKDQRAREGK QFSSADEAAL KEPIIKKFEE EGNPYYSSAR VWDDGIIDPA DTRLVLGLSF SAALNAPIEK TDFGIFRM.
Specifications
Specifications
| Antigen | MCCC2 |
| Applications | ELISA, Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Monoclonal |
| Clone | 2B3 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Mccc2 |
| Gene Accession No. | Q9HCC0 |
| Gene Alias | 3-methylcrotonyl-CoA carboxylase 2; 3-methylcrotonyl-CoA carboxylase non-biotin-containing subunit; 3-methylcrotonyl-CoA:carbon dioxide ligase subunit beta; 4930552N12Rik; biotin carboxylase; MCCase subunit beta; MCCB; Mccc2; meth; methylcrotonoyl-CoA carboxylase 2; methylcrotonoyl-CoA carboxylase 2 (beta); methylcrotonoyl-CoA carboxylase beta chain, mitochondrial; methylcrotonoyl-Coenzyme A carboxylase 2 (beta); non-biotin containing subunit of 3-methylcrotonyl-CoA carboxylase; testicular secretory protein Li 29 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.