Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCCC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584504
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584504 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584504 Supplier Invitrogen™ Supplier No. PA584504
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPCIYLVDSG GAYLPRQADV FPDRDHFGRT FYNQAIMSSK NIAQIAVVMG SCTAGGAYVP AMADENIIVR KQGTIFLAGP Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 93%.

COX3 is a multi-pass membrane protein. It belongs to the cytochrome c oxidase subunit 3 family. Defects in COX3 are a cause of Leber hereditary optic neuropathy (LHON) and cytochrome c oxidase deficiency (COX deficiency). Defects in MT-CO3 are also found in mitochondrial encephalomyopathy with lactic acidosis and stroke-like episodes (MELAS) syndrome and recurrent myoglobinuria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCCC2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mccc2
Gene Accession No. Q9HCC0
Gene Alias 3-methylcrotonyl-CoA carboxylase 2; 3-methylcrotonyl-CoA carboxylase non-biotin-containing subunit; 3-methylcrotonyl-CoA:carbon dioxide ligase subunit beta; 4930552N12Rik; biotin carboxylase; MCCase subunit beta; MCCB; Mccc2; meth; methylcrotonoyl-CoA carboxylase 2; methylcrotonoyl-CoA carboxylase 2 (beta); methylcrotonoyl-CoA carboxylase beta chain, mitochondrial; methylcrotonoyl-Coenzyme A carboxylase 2 (beta); non-biotin containing subunit of 3-methylcrotonyl-CoA carboxylase; testicular secretory protein Li 29
Gene Symbols Mccc2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MCCC2. Recombinant protein control fragment (Product #RP-103413).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64087
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.