Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556803
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556803 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556803 Supplier Invitrogen™ Supplier No. PA556803
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEHGRNELFT DSECKEISEI TGGPINETLK LLLSSSEWVQ SEKLVRALQT PLMRLCAWYL YGEKHRGYAL NPVANFHLQN GAVLWRINWM ADVSLRGITG SCGLMANYRY FLEETGPNST SYLGSKIIKA SEQVLSLVAQ FQKNSKL Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

MCD catalyzes the breakdown of malonyl-CoA to acetyl-CoA and carbon dioxide. Malonyl-CoA is an intermediate in fatty acid biosynthesis, and also inhibits the transport of fatty acyl CoAs into mitochondria. Consequently, the encoded protein acts to increase the rate of fatty acid oxidation. It is found in mitochondria, peroxisomes, and the cytoplasm. Mutations in this gene result in malonyl-CoA decarboyxlase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCD
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MLYCD
Gene Accession No. O95822
Gene Alias AI324784; malonyl coenzyme A decarboxylase; malonyl-CoA decarboxylase; malonyl-CoA decarboxylase, mitochondrial; MCD; MLYCD
Gene Symbols MLYCD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MCD. Recombinant protein control fragment (Product #RP-102339).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23417
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.