Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MCD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597972
Description
MCD Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
MCD catalyzes the breakdown of malonyl-CoA to acetyl-CoA and carbon dioxide. Malonyl-CoA is an intermediate in fatty acid biosynthesis, and also inhibits the transport of fatty acyl CoAs into mitochondria. Consequently, the encoded protein acts to increase the rate of fatty acid oxidation. It is found in mitochondria, peroxisomes, and the cytoplasm. Mutations in this gene result in malonyl-CoA decarboyxlase deficiency.
Specifications
MCD | |
Polyclonal | |
Unconjugated | |
MLYCD | |
AI324784; malonyl coenzyme A decarboxylase; malonyl-CoA decarboxylase; malonyl-CoA decarboxylase, mitochondrial; MCD; MLYCD | |
Rabbit | |
Protein G | |
RUO | |
23417 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O95822 | |
MLYCD | |
Recombinant Human Malonyl-CoA decarboxylase, mitochondrial protein (40-493AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction