Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCEE Monoclonal Antibody (OTI11G7)
GREENER_CHOICE

Catalog No. PIMA527029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527029 Supplier Invitrogen™ Supplier No. MA527029
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MCEE Monoclonal Antibody for Western Blot, IHC (P)

MCEE catalyzes the interconversion of D- and L-methylmalonyl-CoA during the degradation of branched chain amino acids. odd chain-length fatty acids, and other metabolites. Mutations in this gene result in methylmalonyl-CoA epimerase deficiency, which is presented as mild to moderate methylmalonic aciduria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCEE
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI11G7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MCEE
Gene Accession No. Q96PE7
Gene Alias 1110007A04Rik; DL-methylmalonyl-CoA racemase; GLOD2; glyoxalase domain containing 2; Mcee; methylmalonyl CoA epimerase; methylmalonyl-CoA epimerase; methylmalonyl-CoA epimerase, mitochondrial
Gene Symbols MCEE
Host Species Mouse
Immunogen Full length human recombinant protein of MCEE produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84693
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.