Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCEE Monoclonal Antibody (OTI1B5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA808532

Description
MCEE catalyzes the interconversion of D- and L-methylmalonyl-CoA during the degradation of branched chain amino acids. odd chain-length fatty acids, and other metabolites. Mutations in this gene result in methylmalonyl-CoA epimerase deficiency, which is presented as mild to moderate methylmalonic aciduria.Specifications
MCEE | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96PE7 | |
MCEE | |
Full length human recombit protein of human MCEE produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1B5 | |
Unconjugated | |
MCEE | |
GLOD2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
84693 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction