Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCEE Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304474100UL
Description
MCEE Polyclonal specifically detects MCEE in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MCEE | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
84693 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry | |
Unconjugated | |
DL-methylmalonyl-CoA racemase, EC 5.1.99.1, GLOD2, glyoxalase domain containing 2, methylmalonyl CoA epimerase, methylmalonyl-CoA epimerase, mitochondrial | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-176 of human MCEE (NP_115990.3). | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction