Learn More
MCEF Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302538AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene belongs to the AF4 family of transcription factors involved in leukemia. It is a component of the positive transcription elongation factor b (P-TEFb) complex. A chromosomal translocation involving this gene and MLL gene on chromosome 11 is found in infant acute lymphoblastic leukemia with ins.Specifications
MCEF | |
Polyclonal | |
Unconjugated | |
AFF4 | |
AF5Q31, MCEF, AF4/FMR2 family member 4, ALL1-fused gene from chromosome 5q31 protein, major CDK9 elongation factor-associated protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27125 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9UHB7 | |
AFF4 | |
Between 50 and 100 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.