Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCEMP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553223
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553223 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553223 Supplier Invitrogen™ Supplier No. PA553223
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSKELLGFKR ELWNVSNSVQ ACEERQKRGW DSVQQSITMV RSKIDRLETT LAGIKNIDTK VQKILEVLQK MPQSSPQ Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 37%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen MCEMP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MCEMP1
Gene Accession No. Q8IX19
Gene Alias C19orf59; mast cell expressed membrane protein 1; mast cell-expressed membrane protein 1; MCEMP1; MGC132456
Gene Symbols MCEMP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MCEMP1. Recombinant protein control fragment (Product #RP-90532).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199675
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.