Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCEMP1 Rabbit anti-Human, Mouse, Polyclonal, ProteinTech
SDP

Catalog No. 502238493 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-8493 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-8493 Supplier Proteintech Group Inc Supplier No. 257001AP150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Aliquoting is unnecessary for -20°C storage.

MCEMP1 Polyclonal antibody specifically detects MCEMP1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MCEMP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 800 μg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene MCEMP1
Gene Accession No. Q8IX19, Q9D8U6
Gene Alias C19orf59; mast cell expressed membrane protein 1; mast cell-expressed membrane protein 1; MCEMP1; MGC132456
Gene Symbols MCEMP1
Host Species Rabbit
Immunogen C19orf59 Fusion Protein Ag22616
Purification Method Antigen affinity chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199675, 69189
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.