Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588796 Supplier Invitrogen™ Supplier No. PA588796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKMTWLKEIR NILLKQQELL TVKKRKQQDQ LTERDKFQIS LQQNDEKQQG AFISTEETEL EHTSTVVEVC EAIASVQAEA NTVWTEASQS AEISEEPAEW SSNYFYPTYD ENEEENRPLM RPVSEMALLY; Positive Samples: HT-29; Cellular Location: Cytoplasm, Membrane.

Guanine nucleotide exchange factor (GEF) that modulates the Rho family of GTPases. Promotes the conversion of some member of the Rho family GTPase from the GDP-bound to the GTP-bound form. Isoform 1 exhibits no activity toward RHOA, RAC1 or CDC42. Isoform 2 exhibits decreased GEF activity toward CDC42. Isoform 3 exhibits a weak but significant activity toward RAC1 and CDC42. Isoform 4 exhibits significant activity toward RHOA and CDC42. The truncated DBL oncogene is active toward RHOA, RAC1 and CDC42.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCF2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MCF2
Gene Accession No. P10911
Gene Alias ARHGEF21; B230117G22Rik; DBL; DBL-transforming protein; MCF.2 cell line derived transforming sequence; mcf.2 transforming sequence; MCF2; Mcf-2; MCF2-transforming protein; Oncogene MCF2 (oncogene DBL); OTTHUMP00000024159; proto-oncogene DBL; Proto-oncogene MCF-2; RGD1566098
Gene Symbols MCF2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 872-1001 of human MCF2 (NP_0011653471).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109904, 4168
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.