Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCF2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100662
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100662 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100662 Supplier Invitrogen™ Supplier No. PA5100662
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MCF2L.

This gene encodes a guanine nucleotide exchange factor that interacts specifically with the GTP-bound Rac1 and plays a role in the Rho/Rac signaling pathways. A variant in this gene was associated with osteoarthritis. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCF2L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MCF2L
Gene Accession No. O15068, Q63406
Gene Alias ARHGEF14; DBL's big sister; DBS; FLJ12122; Guanine nucleotide exchange factor DBS; KIAA0362; mcf 2l; MCF.2 cell line derived transforming sequence like; MCF.2 cell line derived transforming sequence-like; mcf.2 transforming sequence-like; MCF2 transforming sequence-like protein; MCF2L; MCF2-transforming sequence-like protein; OST; OST oncogene; RP11-98F14.2
Gene Symbols MCF2L
Host Species Rabbit
Immunogen A synthesized peptide derived from human MCF2L(Accession O15068), corresponding to amino acid residues V295-Q345.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117020, 23263
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.