Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCF2L Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | MCF2L |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
MCF2L Polyclonal specifically detects MCF2L in Human, Mouse samples. It is validated for Western Blot.Specifications
MCF2L | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
DBL's big sister, DBS, FLJ12122, guanine nucleotide exchange factor DBS, KIAA0362ARHGEF14, MCF.2 cell line derived transforming sequence-like, MCF2 transforming sequence-like protein, MCF2-transforming sequence-like protein, OST | |
Recombinant fusion protein containing a sequence corresponding to amino acids 850-980 of human MCF2L (NP_079255.4). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis | |
PBS with 50% glycerol, pH7.3. | |
23263 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title