Learn More
mCherry Antibody (1C51), Alexa Fluor™ 700, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP196752AF700
Catalog No. NBP196752Z
Only null left
Add to Cart
Description
Description
mCherry Monoclonal antibody specifically detects mCherry in Nonspecies specific samples. It is validated for Western Blot, Immunohistochemistry, Immunoassay.
Specifications
Specifications
mCherry
Monoclonal
Alexa Fluor 700
DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein
This mCherry Antibody (1C51) was developed against recombinant full-length mCherry purified from E. coli.
Protein G purified
RUO
Primary
Non-species specific, Mouse, Rat, Drosophila, Insect
Purified
Western Blot, Immunohistochemistry, Immunoassay
1C51
Western Blot, Immunohistochemistry, Knockout Validated
Mouse
27 kDa
0.1 mL
Cellular Markers, Fusion Proteins
This mCherry Antibody (1C51) does not cross react with GFP.
Store at 4C in the dark.
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only