Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mCherry Antibody (1C51), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP196752IR
Description
mCherry Monoclonal antibody specifically detects mCherry in Nonspecies specific samples. It is validated for Western Blot, Immunohistochemistry, Immunoassay.Specifications
mCherry | |
Monoclonal | |
DyLight 755 | |
DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein | |
This mCherry Antibody (1C51) was developed against recombinant full-length mCherry purified from E. coli. | |
Protein G purified | |
RUO | |
Primary | |
Non-species specific, Mouse, Rat, Drosophila, Insect | |
Purified |
Western Blot, Immunohistochemistry, Immunoassay | |
1C51 | |
Western Blot, Immunohistochemistry, Knockout Validated | |
Mouse | |
27 kDa | |
0.1 mL | |
Cellular Markers, Fusion Proteins | |
This mCherry Antibody (1C51) does not cross react with GFP. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction