Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mCherry Antibody (1C51), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP196752H
Description
mCherry Monoclonal antibody specifically detects mCherry in Nonspecies specific samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay.Specifications
mCherry | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
Mouse | |
27 kDa | |
0.1 mL | |
Cellular Markers, Fusion Proteins | |
This mCherry Antibody (1C51) does not cross react with GFP. | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay | |
1C51 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated | |
DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein | |
This mCherry Antibody (1C51) was developed against recombinant full-length mCherry purified from E. coli. | |
Protein G purified | |
RUO | |
Primary | |
Non-species specific, Mouse, Rat, Drosophila, Insect | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction