Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mCherry Antibody (1C51), Novus Biologicals™
SDP

Catalog No. NBP196752SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP196752SS 0.025 mL
NBP196752 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP196752SS Supplier Novus Biologicals Supplier No. NBP196752SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 48 publications

mCherry Monoclonal antibody specifically detects mCherry in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, Single Cell Western.

Specifications

Antigen mCherry
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, Western Blot
Classification Monoclonal
Clone 1C51
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:2000, Flow Cytometry, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated, Single Cell Western 100 μg/mL
Formulation 50% PBS, 50% glycerol with 5mM Sodium Azide
Gene Alias DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein
Host Species Mouse
Immunogen This mCherry Antibody (1C51) was developed against recombinant full-length mCherry purified from E. coli.
Molecular Weight of Antigen 27 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Fusion Proteins
Primary or Secondary Primary
Test Specificity This mCherry Antibody (1C51) does not cross react with GFP.
Target Species Non-species specific, Mouse, Rat, Drosophila, Insect
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.