Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mCherry Antibody (235), Novus Biologicals™
SDP

Catalog No. NBP243768 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP243768 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP243768 Supplier Novus Biologicals Supplier No. NBP243768
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot

mCherry Monoclonal specifically detects mCherry in Non-species specific samples. It is validated for Western Blot.

Specifications

Antigen mCherry
Applications Western Blot
Classification Monoclonal
Clone 235
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS with No Preservative
Gene Alias DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein
Host Species Mouse
Immunogen This mCherry Antibody (235) was developed against full length mCherry recombinant protein.
Molecular Weight of Antigen 27 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Fusion Proteins
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.