Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mCherry Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$293.50 - $652.82

Specifications

Antigen mCherry
Concentration 1 mg/mL
Dilution Western Blot 1:1000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin, Live Imaging Microscopy, Immunohistochemistry Whole-Mount, Knockdown Validated, Fluorescence Imaging
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP225157SS
SDP
View Documents
Novus Biologicals
NBP225157SS
0.025 mL
Each for $293.50
Only null left
Add to Cart
 
NBP225157
SDP
View Documents
Novus Biologicals
NBP225157
0.1 mL
Each for $652.82
Only null left
Add to Cart
 
Description

Description

mCherry Polyclonal specifically detects mCherry in Non-species specific samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Live Imaging Microscopy, Knockdown Validated, Fluorescence Imaging.
Specifications

Specifications

mCherry
Western Blot 1:1000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin, Live Imaging Microscopy, Immunohistochemistry Whole-Mount, Knockdown Validated, Fluorescence Imaging
Polyclonal
Rabbit
Cellular Markers, Fusion Proteins
50% PBS, 50% glycerol with 5mM Sodium Azide
This mCherry Antibody was developed against full length recombinant mCherry protein
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
27 kDa
1 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Unconjugated
RUO
Non-species specific, Mouse, Rat, Bacteria
DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein
IgG
Affinity Purified
This antibody detects mCherry.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.