Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mCherry Chicken anti-All, Polyclonal, MilliporeSigma™

Catalog No. AB356481MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB356481MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB356481MI Supplier MilliporeSigma Supplier No. AB356481
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Detect mCherry protein using this chicken polyclonal anti-mCherry Antibody. It has been tested in Immunofluorescence and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mCherry
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols mCherry
Host Species Chicken
Immunogen His-tagged full length recombinant mCherry from Discosoma sp (Mushroom anemone).
Purification Method Unpurified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Secondary and Control Antibodies
Primary or Secondary Primary
Test Specificity This polyclonal antibody detects mCherry in HEK293 cells tranfected with pFin-mCherry.
Target Species All species
Content And Storage Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.