Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mCherry Monoclonal Antibody (OTI10G6), Magnetic beads, TrueMAB™, OriGene
SDP

Catalog No. 502543174
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3174 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3174 Supplier OriGene Supplier No. TA183030S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

mCherry Monoclonal antibody specifically detects mCherry in Tag samples. It is validated for Immunoprecipitation

Specifications

Antigen mCherry
Applications Immunoprecipitation
Classification Monoclonal
Clone OTI10G6
Conjugate Magnetic Beads
Formulation PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide
Gene Alias mCherry
Host Species Mouse
Immunogen Recombinant protein and recognizes tdTomato.
Purification Method Affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.