Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mCherry Mouse anti-All, Clone: 1C51, MilliporeSigma™

Catalog No. MAB131873MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB131873MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB131873MI Supplier MilliporeSigma Supplier No. MAB131873
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-mCherry Antibody, clone IC51.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mCherry
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1C51
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols mCherry
Host Species Mouse
Immunogen His-tagged full length recombinant mCherry.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Secondary and Control Antibodies
Primary or Secondary Primary
Test Specificity Clone 1C51 detects mCherry protein in multiple species.
Target Species All species
Content And Storage Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.